Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 2, 3 | 628 | ||
NM_001322855.1 | NP_001309784.1 | 3, 4 | 770 | |||
NM_001322856.1 | NP_001309785.1 | 3, 4 | 727 | |||
NM_001322857.1 | NP_001309786.1 | 2, 3 | 548 | |||
NM_001322858.1 | NP_001309787.1 | 2, 3 | 495 | |||
NM_001322865.1 | NP_001309794.1 | 2, 3 | 495 | |||
NM_001322868.1 | NP_001309797.1 | 2, 3 | 495 | |||
NM_020448.4 | NP_065181.1 | 2, 3 | 490 | |||
NR_136506.1 | 2, 3 | 495 | ||||
NR_136508.1 | 2, 3 | 495 | ||||
XM_017001869.1 | XP_016857358.1 | 2, 3 | 475 | |||
XM_017001870.1 | XP_016857359.1 | 3, 4 | 740 | |||
GenBank mRNA | AK312433.1 | - | 2, 3 | 444 | ||
BC001265.1 | - | 2, 3 | 432 | 3452234 | ||
BC063583.1 | - | 2, 3 | 420 | 4386115 | ||
U90918.1 | - | 2, 3 | 381 |