Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 3 | 686 | ||
NM_001322855.1 | NP_001309784.1 | 4 | 828 | |||
NM_001322856.1 | NP_001309785.1 | 4 | 785 | |||
NM_001322857.1 | NP_001309786.1 | 3 | 606 | |||
NM_001322858.1 | NP_001309787.1 | 3 | 553 | |||
NM_001322862.1 | NP_001309791.1 | 4 | 663 | |||
NM_001322863.1 | NP_001309792.1 | 3 | 635 | |||
NM_001322864.1 | NP_001309793.1 | 2 | 203 | |||
NM_001322865.1 | NP_001309794.1 | 3 | 553 | |||
NM_001322866.1 | NP_001309795.1 | 3 | 526 | |||
NM_001322868.1 | NP_001309797.1 | 3 | 553 | |||
NM_020448.4 | NP_065181.1 | 3 | 548 | |||
NR_136506.1 | 3 | 553 | ||||
NR_136508.1 | 3 | 553 | ||||
XM_011541808.2 | XP_011540110.1 | 2 | 463 | |||
XM_017001868.1 | XP_016857357.1 | 2 | 198 | |||
XM_017001869.1 | XP_016857358.1 | 3 | 533 | |||
XM_017001870.1 | XP_016857359.1 | 4 | 798 | |||
GenBank mRNA | AK312433.1 | - | 3 | 502 | ||
BC001265.1 | - | 3 | 490 | 3452234 | ||
BC063583.1 | - | 3 | 478 | 4386115 | ||
BX640883.1 | - | 4 | 610 | |||
U90918.1 | - | 3 | 439 |