Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020533.2 | NP_065394.1 | 14 | 1870 | ||
GenBank mRNA | AA831948.1 | - | 1 | 277 | ||
AF171088.1 | - | 5 | 559 | |||
AF249319.1 | - | 14 | 1813 | |||
AF287269.1 | - | 14 | 1854 | |||
AJ293659.1 | - | 13 | 2044 | |||
AJ293970.1 | - | 14 | 1809 | |||
AK026102.1 | - | 14 | 1824 | |||
AK222673.1 | - | 14 | 1849 | |||
AK294330.1 | - | 14 | 1835 | |||
BC005149.2 | - | 14 | 1852 | 3507836 |
Gene Symbol: LOC147791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056629.1 | - | 1 | 1792 | ||
AK125037.1 | - | 1 | 2700 |