Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020533.2 | NP_065394.1 | 4, 5 | 704 | ||
GenBank mRNA | AF249319.1 | - | 4, 5 | 647 | ||
AF287269.1 | - | 4, 5 | 688 | |||
AJ293659.1 | - | 4, 5 | 643 | |||
AJ293970.1 | - | 4, 5 | 643 | |||
AK026102.1 | - | 4, 5 | 658 | |||
AK222673.1 | - | 4, 5 | 683 | |||
AK294330.1 | - | 4, 5 | 669 | |||
BC005149.2 | - | 4, 5 | 686 | 3507836 |