Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020650.2 | NP_065701.2 | 3, 4 | 900 | ||
XM_011527143.1 | XP_011525445.1 | 4, 5 | 1089 | |||
XM_017027023.1 | XP_016882512.1 | 3, 4 | 1174 | |||
GenBank mRNA | AF183423.1 | - | 2, 3 | 485 | ||
AY195859.1 | - | 3, 4 | 900 | |||
AY358423.1 | - | 3, 4 | 487 | |||
BC013436.1 | - | 3, 4 | 483 | 3889718 |