Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305141.1 | NP_001292070.1 | 3 | 454 | ||
NM_001305142.1 | NP_001292071.1 | 3 | 441 | |||
NM_020677.4 | NP_065728.1 | 3 | 555 | |||
XM_006720906.2 | XP_006720969.1 | 4 | 447 | |||
XM_011522567.2 | XP_011520869.1 | 3 | 445 | |||
XM_017023485.1 | XP_016878974.1 | 2 | 418 | |||
XM_017023486.1 | XP_016878975.1 | 2 | 421 | |||
XM_017023487.1 | XP_016878976.1 | 2 | 421 | |||
XM_017023488.1 | XP_016878977.1 | 3 | 530 | |||
XM_017023489.1 | XP_016878978.1 | 4 | 543 | |||
XM_017023491.1 | XP_016878980.1 | 2 | 421 | |||
XM_017023492.1 | XP_016878981.1 | 3 | 530 | |||
GenBank mRNA | AF225419.1 | - | 3 | 560 | ||
AK098784.1 | - | 3 | 415 | |||
BC002927.2 | - | 3 | 335 | 3954035 | ||
BC007364.2 | - | 3 | 397 | 3630659 | ||
HY171572.1 | - | 3 | 456 |