Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AS3MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020682.3 | NP_065733.2 | 9 | 949 | ||
GenBank mRNA | AF226730.1 | - | 9 | 886 | ||
AK057833.1 | - | 9 | 968 | |||
AK225972.1 | - | 9 | 968 | |||
AK309737.1 | - | 9 | 881 | |||
BC119637.1 | - | 9 | 853 | |||
BC119638.2 | - | 9 | 853 |
Gene Symbol: BORCS7-ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037644.1 | 13 | 1214 |