Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022477.3 | NP_071922.2 | 3, 4 | 308 | ||
NM_032013.3 | NP_114402.1 | 4, 5 | 344 | |||
XM_006723837.3 | XP_006723900.1 | 4, 5 | 287 | |||
XM_006723838.3 | XP_006723901.1 | 2, 3 | 389 | |||
XM_011528925.2 | XP_011527227.1 | 2, 3 | 199 | |||
XM_011528926.2 | XP_011527228.1 | 2, 3 | 199 | |||
XM_011528927.2 | XP_011527229.1 | 3, 4 | 251 | |||
XM_017027978.1 | XP_016883467.1 | 4, 5 | 584 | |||
XM_017027979.1 | XP_016883468.1 | 4, 5 | 584 | |||
XM_017027980.1 | XP_016883469.1 | 2, 3 | 358 | |||
GenBank mRNA | AB044943.1 | - | 4, 5 | 284 | ||
AF308609.1 | - | 3, 4 | 206 | |||
BC136489.1 | - | 2, 3 | 225 | 9020478 | ||
DA788570.1 | - | 3, 4 | 303 |