Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020695.3 | NP_065746.3 | 12 | 3245 | ||
XM_011528144.1 | XP_011526446.1 | 12 | 3307 | |||
XM_011528146.2 | XP_011526448.1 | 12 | 3571 | |||
XM_017027028.1 | XP_016882517.1 | 12 | 3280 | |||
XM_017027029.1 | XP_016882518.1 | 12 | 3307 | |||
GenBank mRNA | AB032964.1 | - | 12 | 3233 | ||
AK301030.1 | - | 8 | 928 | |||
AK301729.1 | - | 8 | 1282 | |||
AK302362.1 | - | 9 | 1189 | |||
AK311255.1 | - | 6 | 659 | |||
BC032244.1 | - | 12 | 3255 | 4398472 |