Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145444.1 | NP_001138916.1 | 11 | 1821 | ||
NM_001145445.1 | NP_001138917.1 | 12 | 1866 | |||
NM_020706.2 | NP_065757.1 | 12 | 1866 | |||
XM_005261017.2 | XP_005261074.1 | 11 | 1670 | |||
XM_006724035.2 | XP_006724098.1 | 12 | 1835 | |||
XM_006724036.2 | XP_006724099.1 | 12 | 1835 | |||
XM_017028415.1 | XP_016883904.1 | 12 | 1824 | |||
XM_017028416.1 | XP_016883905.1 | 12 | 1709 | |||
XM_017028417.1 | XP_016883906.1 | 11 | 1670 | |||
GenBank mRNA | AB032998.1 | - | 6 | 854 | ||
AF023142.1 | - | 12 | 1470 | |||
AK025748.1 | - | 7 | 885 | |||
AK308406.1 | - | 11 | 1672 | |||
AL117417.1 | - | 7 | 874 | |||
AL834304.1 | - | 12 | 1646 | |||
BC014921.1 | - | 12 | 1843 | 3907610 | ||
BC043353.1 | - | 12 | 1823 | 6019414 | ||
BC052286.1 | - | 12 | 1697 | 5442227 | ||
BC064990.1 | - | 12 | 1697 | 5432277 |