Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASKIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142643.2 | NP_001136115.1 | 4 | 440 | ||
NM_020753.4 | NP_065804.2 | 5 | 957 | |||
GenBank mRNA | AB032965.1 | - | 1 | 136 | ||
AF451976.1 | - | 5 | 810 | |||
AK300354.1 | - | 4 | 387 | |||
AK315936.1 | - | 4 | 441 | |||
BC066643.1 | - | 5 | 920 | 5576059 | ||
DC318119.1 | - | 4 | 429 |