Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163034.1 | NP_001156506.1 | 9 | 1832 | ||
NM_020761.2 | NP_065812.1 | 9 | 1832 | |||
GenBank mRNA | AB037724.1 | - | 5 | 514 | ||
AB082951.1 | - | 9 | 1027 | |||
AK055912.1 | - | 9 | 1678 | |||
AK296321.1 | - | 9 | 1827 | |||
AY090663.1 | - | 9 | 1389 | |||
BC064515.1 | - | 9 | 1780 | |||
BC136652.1 | - | 9 | 1222 | 9020642 | ||
BC136654.1 | - | 9 | 1222 | 9020644 |