Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020761.2 | NP_065812.1 | 15 | 2405 | ||
GenBank mRNA | AB037724.1 | - | 11 | 1087 | ||
AB082951.1 | - | 15 | 1600 | |||
AK055912.1 | - | 15 | 2251 | |||
AK296321.1 | - | 15 | 2400 | |||
AY090663.1 | - | 15 | 1962 | |||
BC136652.1 | - | 15 | 1795 | 9020642 | ||
BC136654.1 | - | 15 | 1795 | 9020644 |