Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020770.2 | NP_065821.1 | 2, 3 | 1013 | ||
XM_005245365.4 | XP_005245422.1 | 2, 3 | 1025 | |||
GenBank mRNA | AB037740.1 | - | 2, 3 | 986 | ||
AF263462.1 | - | 2, 3 | 1020 | |||
AK290007.1 | - | 2, 3 | 986 | |||
BC146657.1 | - | 2, 3 | 942 | 40147940 | ||
BC150604.1 | - | 1, 2 | 892 | 9056974 | ||
BC152445.1 | - | 2, 3 | 986 | 8862573 |