Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020776.1 | NP_065827.1 | 3, 4 | 249 | ||
NR_136303.1 | 3, 4 | 260 | ||||
NR_136304.1 | 3, 4 | 260 | ||||
XM_005258315.4 | XP_005258372.1 | 3, 4 | 252 | |||
XM_005258317.3 | XP_005258374.1 | 3, 4 | 641 | |||
XM_005258318.4 | XP_005258375.1 | 3, 4 | 256 | |||
XM_011526099.2 | XP_011524401.1 | 3, 4 | 445 | |||
XM_011526100.2 | XP_011524402.1 | 3, 4 | 252 | |||
XM_017025876.1 | XP_016881365.1 | 3, 4 | 562 | |||
XM_017025877.1 | XP_016881366.1 | 3, 4 | 188 | |||
GenBank mRNA | AB037749.2 | - | 3, 4 | 641 | ||
AK292288.1 | - | 3, 4 | 238 |