Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 6, 7 | 1112 | ||
NM_170686.2 | NP_733787.1 | 5, 6 | 1049 | |||
XM_011516439.2 | XP_011514741.1 | 6, 7 | 930 | |||
XM_011516440.2 | XP_011514742.1 | 5, 6 | 902 | |||
XM_011516441.2 | XP_011514743.1 | 5, 6 | 885 | |||
GenBank mRNA | AB037760.1 | - | 1, 2 | 75 | ||
AK290499.1 | - | 5, 6 | 1017 | |||
AK301890.1 | - | 6, 7 | 930 | |||
AK304369.1 | - | 2, 3 | 209 | |||
AK304603.1 | - | 3, 4 | 348 | |||
AY049743.1 | - | 6, 7 | 1112 | |||
AY049744.1 | - | 5, 6 | 791 | |||
BC043295.2 | - | 5, 6 | 1041 | 5298104 |