Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 7 | 1988 | ||
NM_170686.2 | NP_733787.1 | 6 | 1925 | |||
XM_011516439.2 | XP_011514741.1 | 7 | 1806 | |||
XM_011516440.2 | XP_011514742.1 | 6 | 1778 | |||
XM_011516441.2 | XP_011514743.1 | 6 | 1761 | |||
GenBank mRNA | AB037760.1 | - | 2 | 951 | ||
AK290499.1 | - | 6 | 1893 | |||
AK301890.1 | - | 7 | 1806 | |||
AK304369.1 | - | 3 | 1085 | |||
AK304603.1 | - | 4 | 1224 | |||
AY049743.1 | - | 7 | 1988 | |||
AY049744.1 | - | 6 | 1667 | |||
BC043295.2 | - | 6 | 1917 | 5298104 |