Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MICAL3 SIRNA Assays ›
Gene Symbol:
MICAL3
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases:
MICAL-3
Chromosome Location:
Chr.22: 17787649 - 18024559 on Build GRCh38
UniGene ID:
Hs.528024
Species:
Human
siRNA ID
HSS126437
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MICAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122731.2
NP_001116203.1
12, 13
1967
NM_001136004.3
NP_001129476.1
12, 13
1967
NM_015241.2
NP_056056.2
13, 14
2246
XM_005261262.3
XP_005261319.1
13, 14
2076
XM_011546137.2
XP_011544439.1
13, 14
2076
XM_011546139.2
XP_011544441.1
13, 14
2076
XM_011546142.2
XP_011544444.1
13, 14
2076
XM_011546146.2
XP_011544448.1
13, 14
2076
XM_017028865.1
XP_016884354.1
13, 14
2594
XM_017028866.1
XP_016884355.1
13, 14
2400
XM_017028867.1
XP_016884356.1
13, 14
3517
XM_017028868.1
XP_016884357.1
13, 14
2246
XM_017028869.1
XP_016884358.1
13, 14
1991
XM_017028870.1
XP_016884359.1
13, 14
1992
XM_017028871.1
XP_016884360.1
13, 14
2076
XM_017028872.1
XP_016884361.1
13, 14
2076
XM_017028873.1
XP_016884362.1
13, 14
2076
XM_017028874.1
XP_016884363.1
13, 14
2076
XM_017028875.1
XP_016884364.1
13, 14
2076
XM_017028876.1
XP_016884365.1
13, 14
2076
XM_017028877.1
XP_016884366.1
13, 14
2076
XM_017028878.1
XP_016884367.1
13, 14
2076
XM_017028879.1
XP_016884368.1
13, 14
2076
XM_017028880.1
XP_016884369.1
13, 14
2399
XM_017028881.1
XP_016884370.1
13, 14
2246
XM_017028882.1
XP_016884371.1
13, 14
2076
XM_017028883.1
XP_016884372.1
12, 13
1739
XM_017028884.1
XP_016884373.1
14, 15
2525
XM_017028887.1
XP_016884376.1
13, 14
2076
GenBank mRNA
AB037785.1
-
10, 11
1398
AK127487.1
-
12, 13
1975
BC157876.1
-
12, 13
1955
8862700
BC171887.1
-
12, 13
1955
9054541
BX647382.1
-
12, 13
1916
CR456364.1
-
12, 13
1908
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
cytoskeleton organization
actin filament depolymerization
oxidation-reduction process
actin binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
Rab GTPase binding
FAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline