Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020795.3 | NP_065846.1 | 3 | 656 | ||
XM_005256744.3 | XP_005256801.1 | 2 | 606 | |||
XM_017024897.1 | XP_016880386.1 | 3 | 245 | |||
GenBank mRNA | AF376802.1 | - | 3 | 656 |