Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020795.3 | NP_065846.1 | 5 | 964 | ||
XM_005256744.3 | XP_005256801.1 | 4 | 914 | |||
XM_017024897.1 | XP_016880386.1 | 5 | 553 | |||
GenBank mRNA | AB037787.1 | - | 1 | 38 | ||
AF376802.1 | - | 5 | 964 |