Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292003.1 | NP_001278932.1 | 3 | 809 | ||
NM_020803.4 | NP_065854.3 | 3 | 766 | |||
GenBank mRNA | AB037799.2 | - | 2 | 449 | ||
AK128827.1 | - | 3 | 809 | |||
BC041384.2 | - | 3 | 716 | 5275946 | ||
BX538173.1 | - | 3 | 683 | |||
BX640727.1 | - | 3 | 676 | |||
BX647454.1 | - | 3 | 701 |