Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020820.3 | NP_065871.2 | 33 | 4262 | ||
XM_011528934.2 | XP_011527236.1 | 25 | 3164 | |||
GenBank mRNA | AB037836.2 | - | 33 | 4125 | ||
AJ320261.1 | - | 33 | 4262 | |||
AL136579.1 | - | 16 | 2219 | |||
AL832913.1 | - | 26 | 3231 | |||
BC053616.1 | - | 29 | 3712 |