Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB040907.1 | - | 2 | 3018 | ||
AK291466.1 | - | 2 | 3040 | |||
AK293267.1 | - | 1 | 1778 | |||
AL136805.1 | - | 1 | 2499 | |||
BC127095.1 | - | 1 | 2499 | 40123924 | ||
BC127096.1 | - | 1 | 2499 | 40123925 | ||
BX648745.1 | - | 2 | 3041 |