Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 5 | 2445 | ||
XM_005263163.4 | XP_005263220.1 | 4 | 6899 | |||
XM_005263164.4 | XP_005263221.1 | 4 | 1457 | |||
XM_005263165.4 | XP_005263222.1 | 4 | 2883 | |||
XM_006714282.3 | XP_006714345.1 | 5 | 1600 | |||
XM_011532157.2 | XP_011530459.2 | 4 | 2287 | |||
XM_011532158.2 | XP_011530460.1 | 6 | 2246 | |||
XM_011532159.1 | XP_011530461.1 | 5 | 2435 | |||
XM_011532160.1 | XP_011530462.1 | 3 | 1447 | |||
XM_011532161.2 | XP_011530463.1 | 4 | 1625 | |||
XM_017008488.1 | XP_016863977.1 | 5 | 1603 | |||
XM_017008489.1 | XP_016863978.1 | 5 | 1504 | |||
GenBank mRNA | AB055660.1 | - | 5 | 2446 | ||
AK124415.1 | - | 1 | 84 | |||
AK128677.1 | - | 1 | 947 | |||
AK304103.1 | - | 4 | 1241 |