Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029939.3 | NP_001025110.2 | 7 | 1245 | ||
NM_001167583.2 | NP_001161055.1 | 6 | 1086 | |||
NM_001174157.1 | NP_001167628.1 | 5 | 977 | |||
NM_001174158.1 | NP_001167629.1 | 6 | 1086 | |||
NM_001289394.1 | NP_001276323.1 | 7 | 1322 | |||
NM_020863.3 | NP_065914.2 | 6 | 1163 | |||
NR_110323.1 | 6 | 1163 | ||||
XM_011517203.1 | XP_011515505.1 | 6 | 1060 | |||
XM_011517204.2 | XP_011515506.1 | 5 | 883 | |||
XM_011517206.1 | XP_011515508.1 | 6 | 1084 | |||
XM_017013716.1 | XP_016869205.1 | 5 | 958 | |||
GenBank mRNA | AB040918.1 | - | 4 | 561 | ||
AB167738.1 | - | 6 | 1152 | |||
AB167739.1 | - | 7 | 1311 | |||
AB167740.1 | - | 7 | 1238 | |||
AB167741.1 | - | 6 | 1079 | |||
AK300133.1 | - | 6 | 1075 | |||
AK300324.1 | - | 6 | 1082 | |||
AK300471.1 | - | 6 | 1148 | |||
AL832066.1 | - | 6 | 1163 | |||
BC012989.2 | - | 6 | 1077 | |||
BC025423.2 | - | 4 | 755 | |||
BC098405.1 | - | 6 | 1086 |