Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286725.1 | NP_001273654.1 | 7 | 2390 | ||
NM_020866.2 | NP_065917.1 | 8 | 2573 | |||
XM_017020678.1 | XP_016876167.1 | 8 | 1493 | |||
XM_017020679.1 | XP_016876168.1 | 8 | 2080 | |||
GenBank mRNA | AB040923.1 | - | 8 | 2539 | ||
AF252283.1 | - | 8 | 2407 | |||
AK291435.1 | - | 8 | 2573 | |||
AK295939.1 | - | 7 | 1581 | |||
BC022460.1 | - | 8 | 1817 | 4819077 |