Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020886.3 | NP_065937.1 | 2, 3 | 199 | ||
XM_005271630.2 | XP_005271687.1 | 2, 3 | 195 | |||
XM_005271631.2 | XP_005271688.1 | 2, 3 | 195 | |||
XM_005271632.2 | XP_005271689.1 | 3, 4 | 525 | |||
XM_005271633.4 | XP_005271690.1 | 3, 4 | 253 | |||
XM_005271636.2 | XP_005271693.1 | 2, 3 | 195 | |||
XM_005271637.2 | XP_005271694.1 | 2, 3 | 195 | |||
XM_005271638.2 | XP_005271695.1 | 2, 3 | 195 | |||
XM_005271639.2 | XP_005271696.1 | 2, 3 | 195 | |||
XM_017018057.1 | XP_016873546.1 | 3, 4 | 525 | |||
XM_017018063.1 | XP_016873552.1 | 2, 3 | 149 | |||
XM_017018064.1 | XP_016873553.1 | 1, 2 | 70 | |||
XM_017018065.1 | XP_016873554.1 | 2, 3 | 163 | |||
XM_017018066.1 | XP_016873555.1 | 3, 4 | 525 | |||
GenBank mRNA | AF266283.1 | - | 2, 3 | 136 | ||
AK097623.1 | - | 2, 3 | 195 | |||
AK304376.1 | - | 2, 3 | 162 | |||
BC065928.1 | - | 2, 3 | 157 | 6140725 | ||
DB061316.1 | - | 2, 3 | 199 |