Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020904.2 | NP_065955.2 | 17 | 2352 | ||
XM_005259107.2 | XP_005259164.1 | 16 | 2272 | |||
XM_006723301.2 | XP_006723364.1 | 16 | 2272 | |||
XM_011527157.1 | XP_011525459.1 | 17 | 2359 | |||
XM_011527158.2 | XP_011525460.1 | 16 | 2198 | |||
XM_011527159.1 | XP_011525461.1 | 17 | 2372 | |||
XM_011527160.1 | XP_011525462.1 | 17 | 2359 | |||
XM_011527162.2 | XP_011525464.1 | 10 | 968 | |||
XM_017027037.1 | XP_016882526.1 | 14 | 1636 | |||
GenBank mRNA | AB208908.1 | - | 15 | 3844 | ||
AY007233.1 | - | 17 | 2326 | |||
BC024157.2 | - | 17 | 2332 | 3953478 | ||
BC033832.1 | - | 9 | 802 |