Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020905.3 | NP_065956.1 | 2 | 785 | ||
GenBank mRNA | AF237952.1 | - | 2 | 727 | ||
AK023625.1 | - | 1 | 1112 | |||
AK314994.1 | - | 2 | 745 | |||
AY358511.1 | - | 2 | 728 | |||
BC009830.2 | - | 2 | 685 | 4053893 |
Gene Symbol: NT5C1B-RDH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199103.1 | NP_001186032.1 | 9 | 1718 | ||
NM_001199104.1 | NP_001186033.1 | 11 | 2167 |