Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 65, 66 | 15354 | ||
XM_005257545.4 | XP_005257602.2 | 66, 67 | 15542 | |||
XM_005257546.4 | XP_005257603.2 | 66, 67 | 15446 | |||
XM_006721995.3 | XP_006722058.1 | 66, 67 | 15829 | |||
XM_017024905.1 | XP_016880394.1 | 61, 62 | 14409 | |||
GenBank mRNA | AB046774.1 | - | 25, 26 | 3544 | ||
AB537889.1 | - | 65, 66 | 15347 | |||
AK023871.1 | - | 8, 9 | 1173 | |||
AK025676.1 | - | 19, 20 | 2760 | |||
AK025914.1 | - | 18, 19 | 2528 | |||
AK026038.1 | - | 4, 5 | 540 | |||
AK074030.1 | - | 27, 28 | 3898 | |||
AL161961.1 | - | 3, 4 | 1012 | |||
AL833201.1 | - | 25, 26 | 3572 | |||
BC032220.1 | - | 9, 10 | 1318 | |||
BX640932.1 | - | 31, 32 | 4510 | |||
BX647946.1 | - | 37, 38 | 6145 |