Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135745.1 | NP_001129217.1 | 3 | 426 | ||
NM_020919.3 | NP_065970.2 | 3 | 426 | |||
XM_006712654.2 | XP_006712717.1 | 3 | 1003 | |||
XM_017004569.1 | XP_016860058.1 | 2 | 130 | |||
XM_017004570.1 | XP_016860059.1 | 2 | 130 | |||
XM_017004571.1 | XP_016860060.1 | 2 | 130 | |||
GenBank mRNA | AB046783.2 | - | 2 | 147 | ||
AB053305.1 | - | 3 | 193 | |||
AB053306.1 | - | 3 | 193 | |||
AF391100.1 | - | 3 | 193 | |||
AK023024.1 | - | 3 | 193 | |||
AK291029.1 | - | 3 | 210 | |||
BC029174.1 | - | 3 | 196 | |||
BC071576.1 | - | 3 | 188 | 30347073 | ||
BM560426.1 | - | 3 | 457 | |||
BP348546.1 | - | 3 | 425 | |||
KU178735.1 | - | 2 | 70 | |||
KU178736.1 | - | 2 | 70 |