Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 13 | 1524 | ||
NR_135104.1 | 13 | 1596 | ||||
NR_135105.1 | 13 | 1508 | ||||
XM_017019710.1 | XP_016875199.1 | 13 | 1563 | |||
XM_017019711.1 | XP_016875200.1 | 12 | 1618 | |||
XM_017019712.1 | XP_016875201.1 | 13 | 1416 | |||
XM_017019713.1 | XP_016875202.1 | 13 | 1539 | |||
XM_017019714.1 | XP_016875203.1 | 14 | 1697 | |||
XM_017019715.1 | XP_016875204.1 | 11 | 1264 | |||
XM_017019716.1 | XP_016875205.1 | 11 | 1209 | |||
XM_017019717.1 | XP_016875206.1 | 13 | 1461 | |||
XM_017019718.1 | XP_016875207.1 | 11 | 1879 | |||
XM_017019719.1 | XP_016875208.1 | 13 | 1416 | |||
XM_017019720.1 | XP_016875209.1 | 11 | 2494 | |||
XM_017019721.1 | XP_016875210.1 | 11 | 1226 | |||
GenBank mRNA | AB046815.1 | - | 8 | 3351 | ||
AK074219.1 | - | 11 | 1163 | |||
AK307384.1 | - | 13 | 1500 | |||
AL833756.1 | - | 11 | 2545 | |||
BC030020.2 | - | 13 | 1468 | 4821394 | ||
BC035911.1 | - | 2 | 373 |