Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 3 | 278 | ||
NR_135104.1 | 3 | 278 | ||||
NR_135105.1 | 3 | 278 | ||||
XM_011538608.2 | XP_011536910.1 | 3 | 266 | |||
XM_017019710.1 | XP_016875199.1 | 3 | 263 | |||
XM_017019711.1 | XP_016875200.1 | 2 | 318 | |||
XM_017019712.1 | XP_016875201.1 | 3 | 116 | |||
XM_017019713.1 | XP_016875202.1 | 3 | 293 | |||
XM_017019714.1 | XP_016875203.1 | 4 | 451 | |||
XM_017019715.1 | XP_016875204.1 | 3 | 264 | |||
XM_017019716.1 | XP_016875205.1 | 3 | 263 | |||
XM_017019717.1 | XP_016875206.1 | 3 | 225 | |||
XM_017019718.1 | XP_016875207.1 | 2 | 735 | |||
XM_017019719.1 | XP_016875208.1 | 3 | 234 | |||
XM_017019720.1 | XP_016875209.1 | 3 | 219 | |||
XM_017019721.1 | XP_016875210.1 | 3 | 264 | |||
GenBank mRNA | AK074219.1 | - | 3 | 230 | ||
AK131442.1 | - | 2 | 672 | |||
AK301043.1 | - | 3 | 241 | |||
AK307384.1 | - | 3 | 254 | |||
AL833756.1 | - | 3 | 223 | |||
BC030020.2 | - | 3 | 222 | 4821394 | ||
DA806426.1 | - | 3 | 279 |