Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020944.2 | NP_065995.1 | 12 | 2397 | ||
XM_006716809.3 | XP_006716872.1 | 12 | 2442 | |||
XM_011517973.2 | XP_011516275.1 | 12 | 2002 | |||
XM_017014938.1 | XP_016870427.1 | 12 | 2442 | |||
XM_017014940.1 | XP_016870429.1 | 11 | 1783 | |||
XM_017014941.1 | XP_016870430.1 | 11 | 2205 | |||
XM_017014942.1 | XP_016870431.1 | 12 | 2015 | |||
XM_017014943.1 | XP_016870432.1 | 12 | 1997 | |||
XM_017014944.1 | XP_016870433.1 | 11 | 1460 | |||
XM_017014945.1 | XP_016870434.1 | 11 | 1442 | |||
XM_017014946.1 | XP_016870435.1 | 13 | 2053 | |||
GenBank mRNA | AB046825.1 | - | 12 | 2387 | ||
AF258662.1 | - | 9 | 1445 | |||
AJ309567.1 | - | 12 | 2397 | |||
AK027884.1 | - | 12 | 2060 | |||
AK295967.1 | - | 13 | 2390 | |||
AK297435.1 | - | 10 | 1404 | |||
AL834306.1 | - | 12 | 2157 | |||
BC011363.1 | - | 12 | 2356 | 4339156 |