Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020947.3 | NP_065998.3 | 7 | 1204 | ||
XM_005256074.3 | XP_005256131.1 | 7 | 1274 | |||
XM_005256075.2 | XP_005256132.1 | 8 | 1402 | |||
XM_006721240.2 | XP_006721303.1 | 8 | 1357 | |||
XM_011523248.2 | XP_011521550.1 | 9 | 1501 | |||
XM_011523249.2 | XP_011521551.1 | 8 | 1352 | |||
XM_017023510.1 | XP_016878999.1 | 9 | 1521 | |||
XM_017023511.1 | XP_016879000.1 | 7 | 1276 | |||
XM_017023512.1 | XP_016879001.1 | 8 | 1444 | |||
GenBank mRNA | AB046829.2 | - | 1149 | |||
AK291237.1 | - | 7 | 1204 | |||
AK297241.1 | - | 3 | 338 | |||
AK300132.1 | - | 8 | 1441 | |||
AL137316.1 | - | 2 | 73 | |||
BC023251.1 | - | 6 | 1136 | 4470095 | ||
BC060844.1 | - | 7 | 1191 | 30342219 | ||
BM674034.1 | - | 2 | 575 | |||
BM676714.1 | - | 2 | 564 | |||
BM715013.1 | - | 2 | 47 | |||
BM715579.1 | - | 3 | 361 |