Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020947.3 | NP_065998.3 | 5 | 735 | ||
XM_005256074.3 | XP_005256131.1 | 5 | 805 | |||
XM_005256075.2 | XP_005256132.1 | 6 | 933 | |||
XM_006721240.2 | XP_006721303.1 | 6 | 888 | |||
XM_011523248.2 | XP_011521550.1 | 7 | 1032 | |||
XM_011523249.2 | XP_011521551.1 | 6 | 883 | |||
XM_017023510.1 | XP_016878999.1 | 7 | 1052 | |||
XM_017023511.1 | XP_016879000.1 | 5 | 807 | |||
XM_017023512.1 | XP_016879001.1 | 6 | 975 | |||
GenBank mRNA | AB046829.2 | - | 680 | |||
AK021995.1 | - | 1 | 1476 | |||
AK291237.1 | - | 5 | 735 | |||
AK300132.1 | - | 6 | 972 | |||
AK310887.1 | - | 5 | 811 | |||
BC023251.1 | - | 4 | 667 | 4470095 | ||
BC060844.1 | - | 5 | 722 | 30342219 | ||
BI767086.1 | - | 5 | 696 | |||
BU955954.1 | - | 2 | 229 |