Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020949.2 | NP_066000.2 | 7 | 1938 | ||
GenBank mRNA | AB046833.1 | - | 6 | 1389 | ||
AK122655.1 | - | 7 | 1933 | |||
BC022968.1 | - | 6 | 1554 |