Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020961.3 | NP_066012.1 | 10 | 1080 | ||
GenBank mRNA | AB046847.1 | - | 10 | 1035 | ||
AK055555.1 | - | 10 | 1058 | |||
AK296056.1 | - | 8 | 700 | |||
BC006565.2 | - | 10 | 1031 | 3010654 | ||
BC007449.2 | - | 10 | 1031 | 3010654 |