Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172656.1 | NP_001166127.1 | 9 | 2467 | ||
NM_001172659.1 | NP_001166130.1 | 10 | 2590 | |||
NM_020972.2 | NP_066023.2 | 10 | 2557 | |||
XM_006713900.3 | XP_006713963.1 | 10 | 2611 | |||
XM_006713902.3 | XP_006713965.1 | 10 | 2612 | |||
XM_011513540.2 | XP_011511842.1 | 8 | 2025 | |||
XM_011513541.2 | XP_011511843.1 | 10 | 2675 | |||
XM_017008502.1 | XP_016863991.1 | 10 | 4050 | |||
XM_017008503.1 | XP_016863992.1 | 10 | 2612 | |||
XM_017008504.1 | XP_016863993.1 | 10 | 2611 | |||
XM_017008505.1 | XP_016863994.1 | 9 | 4155 | |||
XM_017008510.1 | XP_016863999.1 | 10 | 2520 | |||
GenBank mRNA | AB046863.1 | - | 10 | 2556 | ||
AK126692.1 | - | 9 | 3422 | |||
AK293775.1 | - | 7 | 1950 | |||
AK294710.1 | - | 10 | 2294 | |||
AK294874.1 | - | 10 | 2518 | |||
AK316484.1 | - | 10 | 2590 | |||
BC041866.1 | - | 4 | 2258 | |||
BC137309.1 | - | 11 | 2300 | 9021306 | ||
BC137310.1 | - | 11 | 2300 | 9021307 |