Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021165.3 | NP_066988.1 | 8 | 2508 | ||
XM_005245379.1 | XP_005245436.1 | 9 | 3088 | |||
XM_011509826.1 | XP_011508128.1 | 6 | 2335 | |||
GenBank mRNA | AB051534.1 | - | 8 | 2501 | ||
AB161694.1 | - | 8 | 2501 | |||
AF131833.1 | - | 1 | 356 | |||
AK098386.1 | - | 1 | 715 | |||
AK123336.1 | - | 5 | 1943 | |||
AK296524.1 | - | 9 | 2584 | |||
AL035289.1 | - | 7 | 2082 | |||
BC028036.1 | - | 8 | 2400 | 5242899 |