Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282588.1 | NP_001269517.1 | 2 | 524 | ||
NM_001282589.1 | NP_001269518.1 | 2 | 524 | |||
NM_001282592.1 | NP_001269521.1 | 2 | 524 | |||
NM_021181.4 | NP_067004.3 | 2 | 524 | |||
XM_011509828.1 | XP_011508130.1 | 2 | 334 | |||
XM_011509829.1 | XP_011508131.1 | 2 | 334 | |||
GenBank mRNA | AB027233.1 | - | 2 | 294 | ||
AF291815.1 | - | 2 | 290 | |||
AF390894.1 | - | 2 | 299 | |||
AJ271869.1 | - | 2 | 312 | |||
AJ276429.2 | - | 2 | 312 | |||
AK290706.1 | - | 2 | 308 | |||
AK292148.1 | - | 2 | 308 | |||
AK298499.1 | - | 2 | 331 | |||
AK298548.1 | - | 2 | 332 | |||
AY358512.1 | - | 2 | 316 | |||
BC027867.1 | - | 2 | 290 | 5203638 | ||
DC388332.1 | - | 2 | 506 |