Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282590.1 | NP_001269519.1 | 3 | 638 | ||
NM_001282592.1 | NP_001269521.1 | 4 | 959 | |||
NM_001282594.1 | NP_001269523.1 | 3 | 677 | |||
NM_001282595.1 | NP_001269524.1 | 3 | 420 | |||
NM_001282596.1 | NP_001269525.1 | 3 | 638 | |||
NM_021181.4 | NP_067004.3 | 4 | 959 | |||
XM_011509828.1 | XP_011508130.1 | 4 | 808 | |||
XM_011509829.1 | XP_011508131.1 | 4 | 808 | |||
GenBank mRNA | AB027233.1 | - | 4 | 729 | ||
AF291815.1 | - | 4 | 725 | |||
AF390894.1 | - | 4 | 734 | |||
AJ271869.1 | - | 4 | 747 | |||
AJ276429.2 | - | 4 | 747 | |||
AK290706.1 | - | 4 | 743 | |||
AK292097.1 | - | 3 | 422 | |||
AK292148.1 | - | 4 | 743 | |||
AK301432.1 | - | 3 | 463 | |||
AL834424.1 | - | 3 | 427 | |||
AY358512.1 | - | 4 | 751 | |||
BC027867.1 | - | 4 | 725 | 5203638 | ||
DA644251.1 | - | 3 | 359 | |||
DA978435.1 | - | 3 | 421 | |||
DA978525.1 | - | 3 | 401 |