Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271186.1 | NP_001258115.1 | 4 | 1102 | ||
NM_001271187.1 | NP_001258116.1 | 4 | 437 | |||
NM_021183.4 | NP_067006.3 | 2 | 1030 | |||
NR_073150.1 | 4 | 437 | ||||
XM_006724775.2 | XP_006724838.1 | 3 | 760 | |||
XM_011531376.1 | XP_011529678.1 | 4 | 1170 | |||
GenBank mRNA | AK124801.1 | - | 1 | 340 | ||
AK310997.1 | - | 4 | 404 | |||
AL049685.2 | - | 1 | 262 | |||
AL832461.1 | - | 1 | 581 | |||
AY298955.1 | - | 2 | 1030 | |||
BC003403.1 | - | 1 | 316 | 3449262 | ||
BC051467.1 | - | 2 | 1017 | |||
BI767215.1 | - | 3 | 325 | |||
BX399437.2 | - | 4 | 471 |