Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021196.3 | NP_067019.3 | 18 | 2567 | ||
NM_133478.2 | NP_597812.1 | 23 | 2963 | |||
GenBank mRNA | AB209752.1 | - | 19 | 3370 | ||
AF243499.2 | - | 18 | 2565 | |||
AF293337.2 | - | 22 | 2912 | |||
AF293338.2 | - | 18 | 2565 | |||
AF452248.1 | - | 20 | 2746 | |||
AF453528.1 | - | 19 | 2640 | |||
BC109221.2 | - | 17 | 2308 | 40008543 |