Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 6 | 937 | ||
NM_001242926.1 | NP_001229855.1 | 5 | 886 | |||
NM_001242927.1 | NP_001229856.1 | 4 | 667 | |||
NM_001242928.1 | NP_001229857.1 | 5 | 886 | |||
NM_021188.2 | NP_067011.1 | 5 | 886 | |||
NR_040251.1 | 5 | 854 | ||||
GenBank mRNA | AK223261.1 | - | 5 | 642 | ||
AK293350.1 | - | 6 | 693 | |||
AK298307.1 | - | 6 | 609 | |||
AK298520.1 | - | 5 | 662 | |||
AK299138.1 | - | 4 | 665 | |||
AK301382.1 | - | 5 | 539 | |||
AK303834.1 | - | 5 | 552 | |||
AK309957.1 | - | 6 | 682 | |||
AK310031.1 | - | 4 | 428 | |||
AK315206.1 | - | 5 | 644 | |||
BC004357.1 | - | 5 | 630 | |||
BC050683.1 | - | 5 | 696 | 5752452 | ||
U90919.1 | - | 5 | 618 |