Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127173.1 | NP_001120645.1 | 2, 3 | 370 | ||
NM_021189.3 | NP_067012.1 | 3, 4 | 509 | |||
GenBank mRNA | AF062733.2 | - | 3, 4 | 466 | ||
AF363367.1 | - | 2, 3 | 368 | |||
AF529206.1 | - | 2, 3 | 235 | |||
AK289541.1 | - | 3, 4 | 509 | |||
AK308915.1 | - | 3, 4 | 481 | |||
AY046418.1 | - | 2, 3 | 273 | |||
AY358332.1 | - | 2, 3 | 374 | |||
BC033819.1 | - | 2, 3 | 318 | 5199627 |