Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292017.1 | NP_001278946.1 | 4 | 795 | ||
NM_021204.4 | NP_067027.1 | 4 | 824 | |||
NR_120457.1 | 3 | 592 | ||||
GenBank mRNA | AF087881.1 | - | 4 | 694 | ||
AF113125.1 | - | 4 | 695 | |||
AF177286.1 | - | 2 | 261 | |||
AK022656.1 | - | 4 | 690 | |||
AL518796.3 | - | 4 | 627 | |||
BC001317.1 | - | 3 | 410 | |||
BC065815.1 | - | 4 | 662 | 6526329 | ||
CR457141.1 | - | 4 | 474 |