Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021217.2 | NP_067040.1 | 4 | 974 | ||
GenBank mRNA | AK290979.1 | - | 4 | 910 | ||
AL365370.1 | - | 1 | 38 | |||
BC043354.1 | - | 4 | 935 | 6014754 | ||
BC062561.1 | - | 4 | 936 | 6644579 | ||
X65230.1 | - | 2 | 566 |