Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021218.1 | NP_067041.1 | 4 | 364 | ||
GenBank mRNA | AF161409.1 | - | 4 | 332 | ||
AF161411.2 | - | 4 | 297 | |||
AK000637.1 | - | 4 | 367 | |||
BC014881.1 | - | 4 | 364 | 3907074 | ||
BC065210.1 | - | 4 | 362 | 3456693 |