Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109689.3 | NP_001103159.1 | 6 | 515 | ||
NM_021061.4 | NP_066405.1 | 6 | 530 | |||
XM_005272327.1 | XP_005272384.1 | 6 | 542 | |||
XM_005272328.3 | XP_005272385.1 | 6 | 1015 | |||
XM_006716612.3 | XP_006716675.1 | 6 | 1030 | |||
XM_006716613.3 | XP_006716676.1 | 6 | 569 | |||
XM_011517209.1 | XP_011515511.1 | 6 | 542 | |||
GenBank mRNA | AB208810.1 | - | 6 | 485 | ||
AK054888.1 | - | 6 | 530 | |||
AK095705.1 | - | 6 | 473 | |||
BC017091.1 | - | 6 | 512 | 3851128 | ||
DB206757.1 | - | 6 | 515 | |||
DB272357.1 | - | 6 | 491 |